4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

Lieferant: Apollo Scientific

OR15225-10G OR15225-1G OR15225-5G
APOSOR15225-10GEA 1690 EUR
APOSOR15225-10G APOSOR15225-1G APOSOR15225-5G
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid
Formel: C₁₀H₉NO₃S
Molecular Weight: 223.25 g/mol
Schmelzpunkt: >256 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01569174
CAS-Nummer: 272437-91-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR