tert-Butyl-3-benzyl-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: N'-Benzylsulphamide, N-BOC protected

OR15251-1G OR15251-500MG OR15251-5G
APOSOR15251-1GEA 159 EUR
APOSOR15251-1G APOSOR15251-500MG APOSOR15251-5G
tert-Butyl-3-benzyl-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate 95%
tert-Butyl-3-benzyl-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate
Formel: C₁₂H₁₈N₂O₄S
Molecular Weight: 286,35 g/mol
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09864818
CAS-Nummer: 147000-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR