2-(Chloromethyl)-1-methyl-1H-imidazole
Lieferant: Apollo Scientific
|
Achtung
|
OR15254-100MG
APOSOR15254-100MGEA
21.9
EUR
APOSOR15254-100MG
2-(Chloromethyl)-1-methyl-1H-imidazole
2-(Chloromethyl)-1-methyl-1H-imidazole
Formel:
C₅H₇ClN₂ Schmelzpunkt: 182…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234119 CAS-Nummer: 19225-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...