3,6-Dichloro[1,2,4]triazolo[4,3-b]pyridazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Dichloro-[1,2,4]triazolo[4,3-b]pyridazine
OR15258-1G
OR15258-250MG
APOSOR15258-1GEA
48
EUR
APOSOR15258-1G
APOSOR15258-250MG
3,6-Dichloro[1,2,4]triazolo[4,3-b]pyridazine
3,6-Dichloro[1,2,4]triazolo[4,3-b]pyridazine
Formel:
C₅H₂Cl₂N₄ Molecular Weight: 189 g/mol Schmelzpunkt: 140…143 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00139673 CAS-Nummer: 33050-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...