4-(2-Propynyloxy)benzenecarbaldehyde 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Prop-2-yn-1-yloxy)benzaldehyde, 4-(2-Propynyloxy)benzaldehyde, 4-(Propargyloxy)benzaldehyde
OR15265-1G
OR15265-5G
OR15265-25G
APOSOR15265-1GEA
21.9
EUR
APOSOR15265-1G
APOSOR15265-5G
APOSOR15265-25G
4-(2-Propynyloxy)benzenecarbaldehyde 99%
4-(2-Propynyloxy)benzenecarbaldehyde
Formel:
C₁₀H₈O₂ Molecular Weight: 160,17 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01088150 CAS-Nummer: 5651-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...