5-Bromo-2-chloro-1H-1,3-benzimidazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-chloro-1H-benzo[d]imidazole
OR15270-1G
OR15270-5G
OR15270-10G
APOSOR15270-1GEA
42.2
EUR
APOSOR15270-1G
APOSOR15270-5G
APOSOR15270-10G
5-Bromo-2-chloro-1H-1,3-benzimidazole 98%
5-Bromo-2-chloro-1H-1,3-benzimidazole
Formel:
C₇H₄BrClN₂ Molecular Weight: 231,49 g/mol Schmelzpunkt: 212…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04128987 CAS-Nummer: 683240-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...