2-(tert-Butyl)-5-methyl-1H-indole 95+%

Lieferant: Apollo Scientific

OR15277-1G OR15277-5G OR15277-10G
APOSOR15277-1GEA 265 EUR
APOSOR15277-1G APOSOR15277-5G APOSOR15277-10G
2-(tert-Butyl)-5-methyl-1H-indole 95+%
2-(tert-Butyl)-5-methyl-1H-indole
Formel: C₁₃H₁₇N
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01463568
CAS-Nummer: 69622-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR