Methyl-2-(1,3-benzoxazol-5-yl)acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-(benzo[d]oxazol-5-yl)acetate
OR15291-100MG
OR15291-500MG
APOSOR15291-100MGEA
33.5
EUR
APOSOR15291-100MG
APOSOR15291-500MG
Methyl-2-(1,3-benzoxazol-5-yl)acetate
Methyl-2-(1,3-benzoxazol-5-yl)acetate
Formel:
C₁₀H₉NO₃ Molecular Weight: 191.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021359 CAS-Nummer: 97479-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...