Methyl-6-chloro-5-iodonicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-chloro-5-iodonicotinate

OR15292-1G OR15292-500MG OR15292-5G
APOSOR15292-1GEA 125 EUR
APOSOR15292-1G APOSOR15292-500MG APOSOR15292-5G
Methyl-6-chloro-5-iodonicotinate
Methyl-6-chloro-5-iodonicotinate
Formel: C₇H₅ClINO₂
Molecular Weight: 297,48 g/mol
Schmelzpunkt: 83…85 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08276963
CAS-Nummer: 365413-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR