Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-(1,3-oxazol-5-yl)benzoate, Methyl 4-(5-Oxazolyl)benzoate

OR15295-1G OR15295-5G
APOSOR15295-1GEA 26.1 EUR
APOSOR15295-1G APOSOR15295-5G
Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate
Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate
Formel: C₁₁H₉NO₃
Molecular Weight: 203.2 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03012557
CAS-Nummer: 179057-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR