3-(3-Bromophenyl)-5-methyl-1H-pyrazole

Lieferant: Apollo Scientific

OR15313-10G OR15313-1G OR15313-5G
APOSOR15313-10GEA 1690 EUR
APOSOR15313-10G APOSOR15313-1G APOSOR15313-5G
3-(3-Bromophenyl)-5-methyl-1H-pyrazole
3-(3-Bromophenyl)-5-methyl-1H-pyrazole
Formel: C₁₀H₉BrN₂
Schmelzpunkt: 101…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276940
CAS-Nummer: 324054-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR