1-[(2-Chloro-1,3-thiazol-5-yl)methyl]piperidine 95%

Lieferant: Apollo Scientific
Achtung

OR15329-250MG
APOSOR15329-250MGEA 50.9 EUR
APOSOR15329-250MG
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]piperidine 95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]piperidine
Formel: C₉H₁₃ClN₂S
Siedepunkt: 102 °C/0,08mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD01443899
CAS-Nummer: 339104-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR