Dimethyl-2-(2-nitrophenyl)malonate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dimethyl 2-(2-nitrophenyl)malonate

OR15334-1G OR15334-5G
APOSOR15334-1GEA 56.7 EUR
APOSOR15334-1G APOSOR15334-5G
Dimethyl-2-(2-nitrophenyl)malonate 95%
Dimethyl-2-(2-nitrophenyl)malonate
Formel: C₁₁H₁₁NO₆
Molecular Weight: 253.21 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03617780
CAS-Nummer: 26465-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR