(3-Methyl-2-furyl)methanol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3-Methylfuran-2-yl)methanol, 2-(Hydroxymethyl)-3-methylfuran
OR15383-1G
OR15383-250MG
APOSOR15383-1GEA
0
EUR
APOSOR15383-1G
APOSOR15383-250MG
(3-Methyl-2-furyl)methanol
(3-Methyl-2-furyl)methanol
Formel:
C₆H₈O₂ Molecular Weight: 112,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817486 CAS-Nummer: 20416-16-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...