6-Chloro-2-(4-hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid
Lieferant: Apollo Scientific
OR15393-10G
OR15393-1G
OR15393-5G
APOSOR15393-10GEA
894
EUR
APOSOR15393-10G
APOSOR15393-1G
APOSOR15393-5G
6-Chloro-2-(4-hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid
6-Chloro-2-(4-hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid
Formel:
C₁₇H₁₂ClNO₃ Molecular Weight: 313.74 g/mol Schmelzpunkt: 350 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200950 CAS-Nummer: 854867-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...