6-Chloro-3-methyl-2-phenyl-4-quinoline carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR15394-10G OR15394-1G OR15394-5G
APOSOR15394-10GEA 894 EUR
APOSOR15394-10G APOSOR15394-1G APOSOR15394-5G
6-Chloro-3-methyl-2-phenyl-4-quinoline carboxylic acid
6-Chloro-3-methyl-2-phenyl-4-quinoline carboxylic acid
Formel: C₁₇H₁₂ClNO₂
Molecular Weight: 297.74 g/mol
Schmelzpunkt: 315 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03419305
CAS-Nummer: 154869-06-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR