5-(4-Chlorophenyl)-2-thiophenecarbaldehyde

Lieferant: Apollo Scientific

OR15397-1G OR15397-5G
APOSOR15397-1GEA 98.6 EUR
APOSOR15397-1G APOSOR15397-5G
5-(4-Chlorophenyl)-2-thiophenecarbaldehyde
5-(4-Chlorophenyl)-2-thiophenecarbaldehyde
Formel: C₁₁H₇ClOS
Molecular Weight: 222,69 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01860767
CAS-Nummer: 38401-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR