Ethyl-2-(5-bromo-1H-indol-3-yl)-2-oxoacetate 95%

Lieferant: Apollo Scientific
Achtung

OR15438-10G OR15438-1G OR15438-5G
APOSOR15438-10GEA 894 EUR
APOSOR15438-10G APOSOR15438-1G APOSOR15438-5G
Ethyl-2-(5-bromo-1H-indol-3-yl)-2-oxoacetate 95%
Ethyl-2-(5-bromo-1H-indol-3-yl)-2-oxoacetate
Formel: C₁₂H₁₀BrNO₃
Molecular Weight: 296,13 g/mol
Schmelzpunkt: 248…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02657946
CAS-Nummer: 17826-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR