4-Propionyl-1H-pyrrole-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR15460-1G
APOSOR15460-1GEA
243
EUR
APOSOR15460-1G
4-Propionyl-1H-pyrrole-2-carboxylic acid
4-Propionyl-1H-pyrrole-2-carboxylic acid
Formel:
C₈H₉NO₃ Molecular Weight: 167,17 g/mol Schmelzpunkt: 216…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200986 CAS-Nummer: 111468-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...