2-[4-{4-[(Benzyloxy)carbonyl]piperazino}-5-chloro-6-oxo-1(6H)-pyridazinyl]acetic acid
Lieferant: Apollo Scientific
Synonyme:
[4-Piperazin-1-yl-5-chloro-6-oxo-6H-pyridazin-1-yl]acetic acid, N4-CBZ protected
OR15475-1G
OR15475-5G
APOSOR15475-1GEA
336
EUR
APOSOR15475-1G
APOSOR15475-5G
2-[4-{4-[(Benzyloxy)carbonyl]piperazino}-5-chloro-6-oxo-1(6H)-pyridazinyl]acetic acid
2-[4-{4-[(Benzyloxy)carbonyl]piperazino}-5-chloro-6-oxo-1(6H)-pyridazinyl]acetic acid
Formel:
C₁₈H₁₉ClN₄O₅ Molecular Weight: 406,83 g/mol Schmelzpunkt: 129…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864830 CAS-Nummer: 1000018-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...