2-Benzyl-1,3-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR15477-1G OR15477-500MG OR15477-5G
APOSOR15477-1GEA 149 EUR
APOSOR15477-1G APOSOR15477-500MG APOSOR15477-5G
2-Benzyl-1,3-thiazole-4-carboxylic acid
2-Benzyl-1,3-thiazole-4-carboxylic acid
Formel: C₁₁H₉NO₂S
Schmelzpunkt: 173…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02102513
CAS-Nummer: 36916-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR