(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-(hydroxymethyl)-1-methyl-1H-imidazole
OR15479-1G
OR15479-500MG
OR15479-100MG
APOSOR15479-1GEA
279
EUR
APOSOR15479-1G
APOSOR15479-500MG
APOSOR15479-100MG
(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol 97%
(5-Bromo-1-methyl-1H-imidazol-2-yl)methanol
Formel:
C₅H₇BrN₂O Molecular Weight: 191.03 g/mol Schmelzpunkt: 143…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864833 CAS-Nummer: 861362-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...