3-[(Dimethylamino)sulphonyl]benzenecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(N,N-Dimethylsulphamoyl)benzoic acid
OR15491-1G
OR15491-25G
OR15491-5G
APOSOR15491-1GEA
66.7
EUR
APOSOR15491-1G
APOSOR15491-25G
APOSOR15491-5G
3-[(Dimethylamino)sulphonyl]benzenecarboxylic acid
3-[(Dimethylamino)sulphonyl]benzenecarboxylic acid
Formel:
C₉H₁₁NO₄S Molecular Weight: 229,26 g/mol Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02708231 CAS-Nummer: 7326-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...