2-Ethoxy-5-pyrimidinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Ethoxypyrimidin-5-yl)boronic acid
OR15522-5G
APOSOR15522-5GEA
179
EUR
APOSOR15522-5G
2-Ethoxy-5-pyrimidinboronsäure
2-Ethoxy-5-pyrimidineboronic acid
Formel:
C₆H₉BN₂O₃ Molecular Weight: 167,9600 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07375134 CAS-Nummer: 1003043-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...