2-Acetamido-5-bromthiazol

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(5-bromothiazol-2-yl)acetamide

OR15548-1G OR15548-5G OR15548-250MG
APOSOR15548-1GEA 94.4 EUR
APOSOR15548-1G APOSOR15548-5G APOSOR15548-250MG
2-Acetamido-5-bromthiazol
2-Acetamido-5-bromthiazol
Formel: C₅H₅BrN₂OS
Molecular Weight: 221,0800 g/mol
Schmelzpunkt: 198…201 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00017336
CAS-Nummer: 7336-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR