N-(4-Chloromethylthiazol-2-yl)acetamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Acetamido-4-(chloromethyl)-1,3-thiazole, N-(4-(Chloromethyl)thiazol-2-yl)acetamide
OR15549-1G
OR15549-250MG
APOSOR15549-1GEA
116
EUR
APOSOR15549-1G
APOSOR15549-250MG
N-(4-Chloromethylthiazol-2-yl)acetamide
N-(4-Chloromethylthiazol-2-yl)acetamide
Formel:
C₆H₇ClN₂OS Molecular Weight: 190,65 g/mol Schmelzpunkt: 166…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022441 CAS-Nummer: 7460-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...