4-Chloromethylthiazol-2-ylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-(chloromethyl)-1,3-thiazole hydrochloride

OR15553-5G
APOSOR15553-5GEA 0 EUR
APOSOR15553-5G
4-Chloromethylthiazol-2-ylamine hydrochloride
4-Chloromethylthiazol-2-ylamine hydrochloride
Formel: C₄H₆Cl₂N₂S
Molecular Weight: 185.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035215
CAS-Nummer: 59608-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR