1-(Triisopropylsilyl)pyrrole-3-boronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR15596-1G OR15596-5G
APOSOR15596-1GEA 145 EUR
APOSOR15596-1G APOSOR15596-5G
1-(Triisopropylsilyl)pyrrole-3-boronic acid 98%
1-(Triisopropylsilyl)pyrrole-3-boronic acid
Formel: C₁₃H₂₆BNO₂Si
Molecular Weight: 267,25 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01114667
CAS-Nummer: 138900-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR