(R)-(+)-1-Boc-3-methylpiperazin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine
OR15642-25G
OR15642-100G
OR15642-5G
APOSOR15642-25GEA
46.4
EUR
APOSOR15642-25G
APOSOR15642-100G
APOSOR15642-5G
(R)-(+)-1-Boc-3-methylpiperazin
(R)-(+)-1-Boc-3-methylpiperazin
Formel:
C₁₀H₂₀N₂O₂ Molecular Weight: 200,28 g/mol Schmelzpunkt: 40…47 °C Flammpunkt: 106,1 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683205 CAS-Nummer: 163765-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...