(S)-1-Boc-2-ethylpiperazin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2S)-2-Ethylpiperazine, N1-BOC protected, (S)-tert-Butyl 2-ethylpiperazine-1-carboxylate
OR15649-250MG
OR15649-5G
APOSOR15649-250MGEA
37.7
EUR
APOSOR15649-250MG
APOSOR15649-5G
(S)-1-Boc-2-ethylpiperazin
(S)-1-Boc-2-ethylpiperazin
Formel:
C₁₁H₂₂N₂O₂ Molecular Weight: 214,30 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07772100 CAS-Nummer: 325145-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...