(S)-1,4-Diazabicyclo[4.3.0]nonane 95%

Lieferant: Apollo Scientific
Gefahr

OR15657-1G OR15657-250MG
APOSOR15657-1GEA 255 EUR
APOSOR15657-1G APOSOR15657-250MG
(S)-1,4-Diazabicyclo[4.3.0]nonane 95%
(S)-1,4-Diazabicyclo[4.3.0]nonane
Formel: C₇H₁₄N₂
Molecular Weight: 126,20 g/mol
Schmelzpunkt: 180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03787926
CAS-Nummer: 93643-24-4
UN: 3267
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR