1-Cyclopropylpiperazine dihydrochloride

Lieferant: Apollo Scientific
Gefahr

OR15662-1G OR15662-250MG
APOSOR15662-1GEA 45.1 EUR
APOSOR15662-1G APOSOR15662-250MG
1-Cyclopropylpiperazine dihydrochloride
1-Cyclopropylpiperazine dihydrochloride
Formel: C₇H₁₄N₂·2HCl
Molecular Weight: 199.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040810
CAS-Nummer: 139256-79-4
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR