(R)-3-Amino-1-benzylpiperidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR15666-1G
OR15666-5G
OR15666-10G
APOSOR15666-1GEA
50.9
EUR
APOSOR15666-1G
APOSOR15666-5G
APOSOR15666-10G
(R)-3-Amino-1-benzylpiperidine 97%
(R)-3-Amino-1-benzylpiperidine
Formel:
C₁₂H₁₈N₂ Molecular Weight: 190,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093374 CAS-Nummer: 168466-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...