(S)-3-(Boc-amino)piperidin 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate

OR15669-5G OR15669-25G OR15669-1G
APOSOR15669-5GEA 34.8 EUR
APOSOR15669-5G APOSOR15669-25G APOSOR15669-1G
(S)-3-(Boc-amino)piperidin 95+%
(S)-3-(Boc-amino)piperidin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Schmelzpunkt: 122…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093383
CAS-Nummer: 216854-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR