Carbamic acid, N-3-azetidinyl-, phenylmethyl ester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Aminoazetidine, 3-CBZ protected

OR15686-1G OR15686-250MG OR15686-5G
APOSOR15686-1GEA 306 EUR
APOSOR15686-1G APOSOR15686-250MG APOSOR15686-5G
Carbamic acid, N-3-azetidinyl-, phenylmethyl ester 95%
Carbamic acid, N-3-azetidinyl-, phenylmethyl ester
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368913
CAS-Nummer: 914348-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR