1-Boc-azetidine-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Formylazetidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde, tert-Butyl 3-Formylazetidine-1-carboxylate, 1-Boc-azetidine-3-carboxaldehyde

OR15687-5G OR15687-1G
APOSOR15687-5GEA 119 EUR
APOSOR15687-5G APOSOR15687-1G
1-Boc-azetidine-3-carboxaldehyde
1-Boc-azetidine-3-carboxaldehyde
Formel: C₉H₁₅NO₃
Molecular Weight: 185,22 g/mol
Siedepunkt: 77…82 °C/2mmHg
Lagertemperatur: Gefrierschrank
MDL: MFCD06656142
CAS-Nummer: 177947-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR