1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Hydroxy-1,2,3,6-tetrahydropyridine, N-BOC protected, tert-Butyl 5-hydroxy-5,6-dihydropyridine-1(2H)-carboxylate
OR15689-1G
OR15689-5G
APOSOR15689-1GEA
203
EUR
APOSOR15689-1G
APOSOR15689-5G
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine
Formel:
C₁₀H₁₇NO₃ Molecular Weight: 199,25 g/mol Schmelzpunkt: 55…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972246 CAS-Nummer: 224779-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...