2-Chlor-5-iodpyridin 98+%

Lieferant: Apollo Scientific
Achtung

OR15739-100G OR15739-25G
APOSOR15739-100GEA 161 EUR
APOSOR15739-100G APOSOR15739-25G
2-Chlor-5-iodpyridin 98+%
2-Chlor-5-iodpyridin
Formel: C₅H₃ClIN
Molecular Weight: 239.44 g/mol
Schmelzpunkt: 95…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863635
CAS-Nummer: 69045-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR