5,6-Dichlor-1H-indol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,6-Dichloroindole

OR15747-100G OR15747-25G OR15747-5G
APOSOR15747-100GEA 957 EUR
APOSOR15747-100G APOSOR15747-25G APOSOR15747-5G
5,6-Dichlor-1H-indol
5,6-Dichloro-1H-indole
Formel: C₈H₅Cl₂N
Molecular Weight: 186.0400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01075036
CAS-Nummer: 121859-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR