2-Chloro-5-nitropyridin-4-amine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2-chloro-5-nitropyridine

OR15938-1G OR15938-5G OR15938-100MG
APOSOR15938-1GEA 24.8 EUR
APOSOR15938-1G APOSOR15938-5G APOSOR15938-100MG
2-Chloro-5-nitropyridin-4-amine 98%
2-Chloro-5-nitropyridin-4-amine
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173,56 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09033797
CAS-Nummer: 2604-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR