1-Boc-2-methylpiperazin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methylpiperazine, N1-BOC protected, tert-Butyl 2-methylpiperazine-1-carboxylate

OR16016-250MG OR16016-25G OR16016-5G
APOSOR16016-250MGEA 21.9 EUR
APOSOR16016-250MG APOSOR16016-25G APOSOR16016-5G
1-Boc-2-methylpiperazin 95%
1-Boc-2-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,2780 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179168
CAS-Nummer: 120737-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR