1-Boc-4-ethylaminopiperidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-4-Ethylaminopiperidine, 4-(Ethylamino)piperidine, N1-BOC protected
OR16030-5G
APOSOR16030-5GEA
186
EUR
APOSOR16030-5G
1-Boc-4-ethylaminopiperidine
1-Boc-4-ethylaminopiperidine
Formel:
C₁₂H₂₄N₂O₂ Molecular Weight: 228,34 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06808580 CAS-Nummer: 264905-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...