3-Hydroxy-2-nitro-6-picolin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methyl-2-nitro-3-pyridon, 6-Methyl-2-nitropyridin-3-ol, 3-Hydroxy-6-methyl-2-nitropyridin
OR16078-100G
OR16078-5G
OR16078-25G
APOSOR16078-100GEA
644
EUR
APOSOR16078-100G
APOSOR16078-5G
APOSOR16078-25G
3-Hydroxy-2-nitro-6-picolin 95%
3-Hydroxy-2-nitro-6-picolin
Formel:
C₆H₆N₂O₃ Molecular Weight: 154,13 g/mol Schmelzpunkt: 105…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006259 CAS-Nummer: 15128-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...