3-Chlor-5-methoxybenzoesäure

Lieferant: Apollo Scientific
Achtung

OR16145-1G OR16145-25G OR16145-5G
APOSOR16145-1GEA 21.9 EUR
APOSOR16145-1G APOSOR16145-25G APOSOR16145-5G
3-Chlor-5-methoxybenzoesäure
3-Chlor-5-methoxybenzoesäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01317994
CAS-Nummer: 82477-67-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR