3-Cyanbenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

OR16147-25G OR16147-5G
APOSOR16147-25GEA 48 EUR
APOSOR16147-25G APOSOR16147-5G
3-Cyanbenzoesäure 98%
3-Cyanbenzoesäure
Formel: C₈H₅NO₂
Molecular Weight: 147.13 g/mol
Schmelzpunkt: 220…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002486
CAS-Nummer: 1877-72-1
EINECS: 217-511-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR