2-Hydroxy-6-methylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Cresotic acid, 6-Hydroxy-o-toluic acid, 6-Methylsalicylic acid

OR16150-1G OR16150-5G OR16150-10G
APOSOR16150-1GEA 21.9 EUR
APOSOR16150-1G APOSOR16150-5G APOSOR16150-10G
2-Hydroxy-6-methylbenzoic acid 95%
2-Hydroxy-6-methylbenzoic acid
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 170…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01194284
CAS-Nummer: 567-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR