Kaliumethylmalonat

Lieferant: Apollo Scientific
Achtung

Synonyme: Monoethyl malonate potassium salt, Ethyl potassium malonate

OR16162-1G
APOSOR16162-1GEA 0 EUR
APOSOR16162-1G
Kaliumethylmalonat
Kaliumethylmalonat
Formel: C₅H₇KO₄
Molecular Weight: 170,21 g/mol
Schmelzpunkt: 194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035603
CAS-Nummer: 6148-64-7
EINECS: 228-156-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR