6-Brom-4-methoxychinolin

Lieferant: Apollo Scientific

OR16166-1G OR16166-5G
APOSOR16166-1GEA 214 EUR
APOSOR16166-1G APOSOR16166-5G
6-Brom-4-methoxychinolin
6-Bromo-4-methoxyquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,0806 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04036192
CAS-Nummer: 874792-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR