3-Isoxazolcarbonsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoxazole-3-carboxylic Acid
OR16167-25G
APOSOR16167-25GEA
126
EUR
APOSOR16167-25G
3-Isoxazolcarbonsäure 96%
3-Isoxazolecarboxylic acid
Formel:
C₄H₃NO₃ Molecular Weight: 113,07 g/mol Schmelzpunkt: 149…152 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06738796 CAS-Nummer: 3209-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...