[4-(Hydroxymethyl)phenoxy]essigsäure 97%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxymethyl-phenoxyacetic acid, HMP Linker

OR16415-25G OR16415-5G
APOSOR16415-25GEA 40.6 EUR
APOSOR16415-25G APOSOR16415-5G
[4-(Hydroxymethyl)phenoxy]essigsäure 97%
4-(Hydroxymethyl)phenoxyacetic acid
Formel: C₉H₁₀O₄
Molecular Weight: 182,2 g/mol
Schmelzpunkt: 107…114 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00057827
CAS-Nummer: 68858-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR